Gene Transcripts

Gene Symbol
Phf20l1
Entrez Gene ID
239510
Gene Name
PHD finger protein 20-like 1
Gene Aliases
CGI-72, E130113K22Rik
Chromosome Location
Chr. 15: 66577517 - 66647043 on Build GRCm38
UniGene ID
Mm.267473
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081409.1 NP_001074878.1 19 2871
GenBank mRNA AK046670.1 - 2 391
AK053606.1 - 9 1230

View Full Product Details