Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001185083.1 | NP_001172012.1 | 3 | 390 | |
NM_001185084.1 | NP_001172013.1 | 2 | 504 | ||
NM_008387.4 | NP_032413.1 | 3 | 402 | ||
GenBank mRNA | AK007612.1 | - | 3 | 403 | |
AY899305.1 | - | 2 | 469 | ||
BC066208.1 | - | 3 | 411 | ||
BC099934.1 | - | 3 | 375 | 6432976 | |
BC132650.1 | - | 3 | 375 | 40130927 | |
BC132652.1 | - | 3 | 375 | 40130929 | |
BC145554.1 | - | 3 | 363 | 9054118 |