Gene Transcripts

Gene Symbol
Adamtsl5
Entrez Gene ID
66548
Gene Name
ADAMTS-like 5
Gene Aliases
2010109H09Rik, Thsd6
Chromosome Location
Chr. 10: 80339793 - 80349036 on Build GRCm38
UniGene ID
Mm.268527
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285435.1 NP_001272364.1 6 582
GenBank mRNA AK008355.1 - 6 547
AK008364.1 - 6 547
AK008440.1 - 6 546
AK028819.1 - 6 1054
AK029081.1 - 6 523
AK154947.1 - 3 172
AK219627.1 - 1 78
BC034843.1 - 4 435
BC058577.1 - 2 1158
CK624091.1 - 1 87

View Full Product Details