Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177461.1 | NP_803412.1 | 16 | 2849 | |
GenBank mRNA | AK007847.1 | - | 3 | 278 | |
AK076226.1 | - | 3 | 150 | ||
AK076763.1 | - | 10 | 1506 | ||
AK077217.1 | - | 9 | 1677 | ||
AK129419.1 | - | 16 | 2587 | ||
AK147587.1 | - | 16 | 2650 | ||
AK147646.1 | - | 16 | 2862 |