Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001113545.1 | NP_001107017.1 | 7 | 1040 | |
NM_023063.4 | NP_075550.2 | 4 | 912 | ||
GenBank mRNA | AF307844.1 | - | 4 | 835 | |
AF307845.1 | - | 7 | 994 | ||
AK028186.1 | - | 4 | 853 | ||
AK031698.1 | - | 4 | 856 | ||
AK049350.1 | - | 7 | 990 | ||
AK085065.1 | - | 4 | 906 | ||
AK132314.1 | - | 4 | 858 | ||
AK167220.1 | - | 4 | 472 | ||
BC031490.1 | - | 7 | 966 | 3498138 |