Gene Transcripts

Gene Symbol
Rhox2g
Entrez Gene ID
434766
Gene Name
reproductive homeobox 2G
Gene Aliases
EG434766, Rhox2.7, Rhox2f
Chromosome Location
Chr. X: 37639112 - 37643470 on Build GRCm38
UniGene ID
Mm.350894
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114153.1 NP_001107625.1 3 546

View Full Product Details