Gene Transcripts

Gene Symbol
Raet1d
Entrez Gene ID
56554
Gene Name
retinoic acid early transcript delta
Gene Aliases
RAE-1delta
Chromosome Location
Chr. 10: 22361865 - 22374143 on Build GRCm38
UniGene ID
Mm.458004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020030.2 NP_064414.1 219
GenBank mRNA AF257520.1 - 2, 3 221
BC106190.1 - 219 30142928
FJ594066.1 - 421

View Full Product Details