Gene Transcripts

Gene Symbol
Prlh
Entrez Gene ID
623503
Gene Name
prolactin releasing hormone
Gene Aliases
EG623503
Chromosome Location
Chr. 1: 90951256 - 90954369 on Build GRCm38
UniGene ID
Mm.471593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101647.1 NP_001095117.1 2 217
GenBank mRNA FJ424613.1 - 2 217

View Full Product Details