Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023133.1 | NP_075622.1 | 2 | 169 | |
GenBank mRNA | AK012052.1 | - | 2 | 48 | |
AK013524.1 | - | 2 | 49 | ||
AK018725.1 | - | 1 | 28 | ||
AK028244.1 | - | 2 | 45 | ||
AK031403.1 | - | 2 | 46 | ||
BC034506.1 | - | 2 | 40 | 3968247 | |
BC086775.1 | - | 1 | 31 | ||
BC086938.1 | - | 2 | 46 | 30121701 |