Gene Transcripts

Gene Symbol
CAMSAP2
Entrez Gene ID
23271
Gene Name
calmodulin regulated spectrin associated protein family member 2
Gene Aliases
CAMSAP1L1
Chromosome Location
Chr. 1: 200738905 - 200860707 on Build GRCh38
UniGene ID
Hs.23585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297707.1 NP_001284636.1 3, 4 824
NM_001297708.1 NP_001284637.1 3, 4 824
NM_203459.2 NP_982284.1 3, 4 824
XM_005245041.3 XP_005245098.1 3, 4 681
XM_017000799.1 XP_016856288.1 2, 3 1217
GenBank mRNA AB029001.2 - 2, 3 182
AK095640.1 - 3, 4 824
AK126821.1 - 2, 3 196
BC125229.1 - 3, 4 603 40124603
BC125230.1 - 3, 4 603 40124604

View Full Product Details