Gene Transcripts

Gene Symbol
CAMSAP2
Entrez Gene ID
23271
Gene Name
calmodulin regulated spectrin associated protein family member 2
Gene Aliases
CAMSAP1L1
Chromosome Location
Chr. 1: 200738905 - 200860707 on Build GRCh38
UniGene ID
Hs.23585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297707.1 NP_001284636.1 12 2701
NM_001297708.1 NP_001284637.1 11 2620
NM_203459.2 NP_982284.1 11 2668
XM_005245041.3 XP_005245098.1 12 2510
XM_017000799.1 XP_016856288.1 11 3094
GenBank mRNA AB029001.2 - 11 2059
AK095640.1 - 11 2668
AK126821.1 - 11 2073
BC056910.1 - 1 1212
BC065508.1 - 1 1019
BC125229.1 - 12 2480 40124603
BC125230.1 - 11 2399 40124604

View Full Product Details