Gene Transcripts

Gene Symbol
Bhlhe23
Entrez Gene ID
499952
Gene Name
basic helix-loop-helix family, member e23
Gene Aliases
Bhlhb4, RGD1559760
Chromosome Location
Chr. 3: 176279997 - 176282211 on Build Rnor_6.0
UniGene ID
Rn.218533
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109211.1 NP_001102681.1 1 1076

View Full Product Details