Gene Transcripts

Gene Symbol
NCAPD3
Entrez Gene ID
23310
Gene Name
non-SMC condensin II complex subunit D3
Gene Aliases
CAP-D3, CAPD3, hCAP-D3, hHCP-6, hcp-6
Chromosome Location
Chr. 11: 134152442 - 134225443 on Build GRCh38
UniGene ID
Hs.438550
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015261.2 NP_056076.1 1 661
XM_011542719.2 XP_011541021.1 1 85
GenBank mRNA AK124878.1 - 1 661
BC098398.1 - 1 120 30528692
BC144524.1 - 1 345
D29954.1 - 1 83

View Full Product Details