Gene Transcripts

Gene Symbol
Hist2h4
Entrez Gene ID
295277
Gene Name
histone cluster 2, H4
Gene Aliases
Hist4h4
Chromosome Location
Chr. 2: 198411955 - 198412350 on Build Rnor_6.0
UniGene ID
Rn.220155
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123469.1 NP_001116941.1 1 198

View Full Product Details