Gene Transcripts

Gene Symbol
Atp6v0b
Entrez Gene ID
298451
Gene Name
ATPase, H+ transporting, lysosomal V0 subunit B
Gene Aliases
Chromosome Location
Chr. 5: 136718352 - 136719868 on Build Rnor_6.0
UniGene ID
Rn.229006
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106681.1 NP_001100151.1 3 574
GenBank mRNA BC159422.1 - 913

View Full Product Details