Gene Transcripts

Gene Symbol
Adamtsl5
Entrez Gene ID
314626
Gene Name
ADAMTS-like 5
Gene Aliases
Thsd6
Chromosome Location
Chr. 7: 12228723 - 12237382 on Build Rnor_6.0
UniGene ID
Rn.13813
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108071.1 NP_001101541.1 8 952
GenBank mRNA FQ224249.1 - 10 1181

View Full Product Details