Gene Transcripts

Gene Symbol
CBX6
Entrez Gene ID
23466
Gene Name
chromobox 6
Gene Aliases
Chromosome Location
Chr. 22: 38861427 - 38872334 on Build GRCh38
UniGene ID
Hs.592201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303494.1 NP_001290423.1 1, 2 217
NM_014292.4 NP_055107.3 1, 2 217
GenBank mRNA AK094829.1 - 1, 2 132
AK293035.1 - 1, 2 99
BC012111.1 - 1, 2 96 4554483
BC064900.1 - 1, 2 84 6009294
CD625491.1 - 1, 2 136
HY151719.1 - 1, 2 181

View Full Product Details