Gene Transcripts

Gene Symbol
MAPK8IP2
Entrez Gene ID
23542
Gene Name
mitogen-activated protein kinase 8 interacting protein 2
Gene Aliases
IB-2, IB2, JIP2, PRKM8IPL
Chromosome Location
Chr. 22: 50600685 - 50613978 on Build GRCh38
UniGene ID
Hs.558180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012324.4 NP_036456.1 3 381
XM_011530679.2 XP_011528981.1 3 270
XM_011530680.2 XP_011528982.1 3 270
XM_011530681.2 XP_011528983.1 3 270
GenBank mRNA AF136382.1 - 3 364
AF218778.1 - 1 166
AK294265.1 - 3 271
BC009940.2 - 3 274 4100637
DB483256.1 - 3 383

View Full Product Details