Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012118.3 | NP_036250.2 | 3 | 1442 | |
GenBank mRNA | AF183961.1 | - | 3 | 1412 | |
AF199494.1 | - | 1 | 1265 | ||
BC113494.1 | - | 3 | 1306 | 8322546 | |
BC113500.1 | - | 3 | 1306 | 8322552 |