Gene Transcripts

Gene Symbol
HECTD1
Entrez Gene ID
25831
Gene Name
HECT domain E3 ubiquitin protein ligase 1
Gene Aliases
EULIR
Chromosome Location
Chr. 14: 31100115 - 31208060 on Build GRCh38
UniGene ID
Hs.708017
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015382.3 NP_056197.3 16 3045
XM_005267502.2 XP_005267559.2 16 3207
XM_011536619.2 XP_011534921.1 16 2643
XM_011536621.2 XP_011534923.1 16 2643
XM_011536622.1 XP_011534924.1 7 970
XM_017021148.1 XP_016876637.1 15 2537
XM_017021149.1 XP_016876638.1 15 2537
XM_017021150.1 XP_016876639.1 15 2537
XM_017021151.1 XP_016876640.1 16 3239
XM_017021152.1 XP_016876641.1 16 3239
XM_017021153.1 XP_016876642.1 16 2643
GenBank mRNA AB032957.2 - 8 1191
AK095634.1 - 7 938
AY254380.1 - 16 2839

View Full Product Details