Gene Transcripts

Gene Symbol
YIPF3
Entrez Gene ID
25844
Gene Name
Yip1 domain family member 3
Gene Aliases
C6orf109, FinGER3, KLIP1, dJ337H4.3
Chromosome Location
Chr. 6: 43511827 - 43516990 on Build GRCh38
UniGene ID
Hs.440950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015388.3 NP_056203.2 3 547
XM_005248989.4 XP_005249046.1 3 583
XM_005248990.2 XP_005249047.1 3 588
XM_011514458.2 XP_011512760.1 3 690
GenBank mRNA AF162672.1 - 3 462
AK000946.1 - 3 483
AK021433.1 - 459
AK021655.1 - 3 427
AK022757.1 - 3 446
AK298150.1 - 3 583
AL050274.1 - 3 348
BC019297.1 - 3 496 2905449
CR533541.1 - 2 259

View Full Product Details