Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015397.3 | NP_056212.1 | 8 | 1436 | |
XM_011517843.1 | XP_011516145.1 | 8 | 1263 | ||
GenBank mRNA | AB067479.1 | - | 8 | 1356 | |
AF131771.1 | - | 3 | 296 | ||
AK000354.1 | - | 8 | 1214 | ||
AK292935.1 | - | 8 | 1242 | ||
AL117646.1 | - | 5 | 570 | ||
BC008893.2 | - | 7 | 999 | ||
BC063823.1 | - | 8 | 1354 | 6184796 | |
DQ518909.1 | - | 8 | 1356 |