Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100409.2 | NP_001093879.1 | 23 | 4292 | |
NM_015571.3 | NP_056386.2 | 24 | 4313 | ||
XM_006715457.3 | XP_006715520.1 | 25 | 4067 | ||
XM_011535746.2 | XP_011534048.1 | 23 | 3865 | ||
XM_011535747.2 | XP_011534049.1 | 23 | 4216 | ||
XM_017010719.1 | XP_016866208.1 | 24 | 3955 | ||
XM_017010720.1 | XP_016866209.1 | 24 | 3271 | ||
XM_017010721.1 | XP_016866210.1 | 24 | 3279 | ||
XM_017010722.1 | XP_016866211.1 | 25 | 4086 | ||
XM_017010723.1 | XP_016866212.1 | 25 | 4063 | ||
XM_017010724.1 | XP_016866213.1 | 25 | 4438 | ||
XM_017010725.1 | XP_016866214.1 | 22 | 4198 | ||
GenBank mRNA | AB018340.2 | - | 24 | 3341 | |
AF196304.1 | - | 24 | 3297 | ||
AF306508.1 | - | 24 | 3312 | ||
AF307849.1 | - | 24 | 3372 | ||
AK021417.1 | - | 14 | 1716 | ||
AK021949.1 | - | 8 | 1236 | ||
AK126921.1 | - | 23 | 4292 | ||
BC028583.1 | - | 23 | 3499 | 4838569 |