Gene Transcripts

Gene Symbol
ARHGEF26
Entrez Gene ID
26084
Gene Name
Rho guanine nucleotide exchange factor 26
Gene Aliases
CSGEF, HMFN1864, SGEF
Chromosome Location
Chr. 3: 154121003 - 154257827 on Build GRCh38
UniGene ID
Hs.240845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251962.1 NP_001238891.1 14 2671
NM_015595.3 NP_056410.3 14 2598
XM_011512672.1 XP_011510974.1 13 2508
XM_017006160.1 XP_016861649.1 13 1340
GenBank mRNA AF415175.1 - 14 2553
AF415176.1 - 4 523
AK022655.1 - 8 909
AK022884.1 - 11 1185
AK127282.1 - 14 2671
AL117429.1 - 4 409
AY552599.1 - 13 2387
BC016628.1 - 8 817
BC078655.1 - 14 2539 4374088

View Full Product Details