Gene Transcripts

Gene Symbol
OR1L1
Entrez Gene ID
26737
Gene Name
olfactory receptor family 1 subfamily L member 1
Gene Aliases
HG23, OR1L2, OR9-27, OR9-C
Chromosome Location
Chr. 9: 122661716 - 122662648 on Build GRCh38
UniGene ID
Hs.567736
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005236.3 NP_001005236.3 1 558

View Full Product Details