Gene Transcripts

Gene Symbol
AKAP8L
Entrez Gene ID
26993
Gene Name
A-kinase anchoring protein 8 like
Gene Aliases
HA95, HAP95, NAKAP, NAKAP95
Chromosome Location
Chr. 19: 15380048 - 15419121 on Build GRCh38
UniGene ID
Hs.399800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291478.1 NP_001278407.1 12 1534
NM_014371.3 NP_055186.3 12 1717
NR_111971.1 14 1752
XM_005259854.3 XP_005259911.2 13 1522
XM_011527916.2 XP_011526218.1 12 1624
GenBank mRNA AB025905.1 - 12 1564
AB451326.1 - 11 1519
AB451469.1 - 11 1519
AF199414.1 - 11 1519
AJ243467.1 - 12 1565
AK001594.1 - 12 2256
AK094736.1 - 14 1619
AK225046.1 - 12 1584
AK225291.1 - 12 1584
AK295956.1 - 12 1439
BC000713.2 - 12 1569 3504388
BF973758.1 - 5 525

View Full Product Details