Gene Transcripts

Gene Symbol
DEXI
Entrez Gene ID
28955
Gene Name
Dexi homolog
Gene Aliases
MYLE
Chromosome Location
Chr. 16: 10928891 - 10943007 on Build GRCh38
UniGene ID
Hs.592051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014015.3 NP_054734.2 1 531
XM_017023171.1 XP_016878660.1 1 1138
GenBank mRNA AF108145.2 - 1 147
AF129929.1 - 1 43
AK314111.1 - 1 500
BC001083.2 - 1 267 2958243
BC037569.1 - 1 531 5227606
BC052989.1 - 1 264 6194689
BC071683.1 - 1 430 5190179

Gene Symbol
LOC100289656
Entrez Gene ID
100289656
Gene Name
Dexi homolog (mouse) pseudogene
Gene Aliases
Chromosome Location
Chr. 15: 28788243 - 28789392 on Build GRCh38
UniGene ID
Hs.646539
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_036475.2 2 837
GenBank mRNA AK302529.1 - 2 837

View Full Product Details