Gene Transcripts
- Gene Symbol
- SAC3D1
- Entrez Gene ID
- 29901
- Gene Name
- SAC3 domain containing 1
- Gene Aliases
- HSU79266, SHD1
- Chromosome Location
- Chr. 11: 65040904 - 65044828 on Build GRCh38
- UniGene ID
- Hs.23642
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013299.3 |
NP_037431.3
|
2 |
1375 |
|
GenBank mRNA |
U79266.1 |
- |
2 |
1370 |
|
View Full Product Details