Gene Transcripts

Gene Symbol
UBQLN2
Entrez Gene ID
29978
Gene Name
ubiquilin 2
Gene Aliases
ALS15, CHAP1, DSK2, HRIHFB2157, N4BP4, PLIC2
Chromosome Location
Chr. X: 56563593 - 56567010 on Build GRCh38
UniGene ID
Hs.179309
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013444.3 NP_038472.2 1 1926
GenBank mRNA AB015344.1 - 1 997
AF189009.1 - 1 1880
AF293385.1 - 1 1645
AL442081.1 - 1 1539
BC069237.1 - 2 1797 4543266

View Full Product Details