Gene Transcripts

Gene Symbol
APIP
Entrez Gene ID
51074
Gene Name
APAF1 interacting protein
Gene Aliases
APIP2, CGI-29, CGI29, MMRP19, hAPIP
Chromosome Location
Chr. 11: 34882295 - 34916411 on Build GRCh38
UniGene ID
Hs.447794
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015957.3 NP_057041.2 3 319
XM_011520154.2 XP_011518456.1 4 392
XM_017017875.1 XP_016873364.1 4 366
GenBank mRNA AF132963.1 - 3 269
AK225249.1 - 3 251
AK226157.1 - 1 223
AK292648.1 - 3 287
AK302225.1 - 4 339
BC008440.1 - 3 286 4096399
BC009077.1 - 3 233 3875061
BC017594.2 - 3 228 2988353
BP200131.1 - 3 408
DA729016.1 - 3 300

View Full Product Details