Gene Transcripts

Gene Symbol
GOLM1
Entrez Gene ID
51280
Gene Name
golgi membrane protein 1
Gene Aliases
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3
Chromosome Location
Chr. 9: 86026141 - 86100201 on Build GRCh38
UniGene ID
Hs.494337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016548.3 NP_057632.2 10 1366
NM_177937.2 NP_808800.1 10 1358
GenBank mRNA AF236056.1 - 10 1307
AK027717.1 - 10 1329
AK075542.1 - 10 1300
AK075557.1 - 10 1270
AK092943.1 - 11 1246
AY358593.1 - 10 1271
BC001740.1 - 10 1281 3351316
CR457201.1 - 9 1157
EU794616.1 - 10 1325

View Full Product Details