Gene Transcripts

Gene Symbol
MS4A4A
Entrez Gene ID
51338
Gene Name
membrane spanning 4-domains A4A
Gene Aliases
4SPAN1, CD20-L1, CD20L1, HDCME31P, MS4A4, MS4A7
Chromosome Location
Chr. 11: 60280541 - 60308972 on Build GRCh38
UniGene ID
Hs.325960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243266.1 NP_001230195.1 5 551
NM_024021.3 NP_076926.2 7 856
NM_148975.2 NP_683876.1 6 710
GenBank mRNA AB013102.1 - 6 551
AB022821.1 - 6 575
AF068288.1 - 6 644
AF237912.1 - 6 661
AF350500.1 - 5 614
AF354928.1 - 4 375
AK075081.1 - 7 741
AK292089.1 - 6 665
AK314033.1 - 6 553
BC020648.1 - 6 585 4720088
CB046393.1 - 5 420

Gene Symbol
MS4A4E
Entrez Gene ID
643680
Gene Name
membrane spanning 4-domains A4E
Gene Aliases
Chromosome Location
Chr. 11: 60201253 - 60243124 on Build GRCh38
UniGene ID
Hs.559934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011545416.2 XP_011543718.1 6 593
XM_011545418.2 XP_011543720.1 5 529
XM_017018641.1 XP_016874130.1 6 593

View Full Product Details