Gene Transcripts
- Gene Symbol
- NUP54
- Entrez Gene ID
- 53371
- Gene Name
- nucleoporin 54
- Gene Aliases
- Chromosome Location
- Chr. 4: 76114659 - 76148515 on Build GRCh38
- UniGene ID
- Hs.430435
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278603.1 |
NP_001265532.1
|
5, 6 |
907 |
|
NM_017426.3 |
NP_059122.2
|
6, 7 |
1051 |
|
NR_103780.1 |
|
5, 6 |
824 |
|
NR_103781.1 |
|
5, 6 |
824 |
|
NR_103782.1 |
|
6, 7 |
821 |
|
XM_006714237.1 |
XP_006714300.1
|
7, 8 |
1028 |
|
XM_011532033.2 |
XP_011530335.1
|
6, 7 |
994 |
|
XM_011532034.2 |
XP_011530336.1
|
5, 6 |
1294 |
|
XM_017008294.1 |
XP_016863783.1
|
6, 7 |
1438 |
|
XM_017008295.1 |
XP_016863784.1
|
5, 6 |
1211 |
|
GenBank mRNA |
AF157322.1 |
- |
6, 7 |
929 |
|
AK001517.1 |
- |
6, 7 |
933 |
|
AK222752.1 |
- |
6, 7 |
938 |
|
AK223352.1 |
- |
6, 7 |
934 |
|
AK299442.1 |
- |
5, 6 |
706 |
|
AK300036.1 |
- |
5, 6 |
789 |
|
AK307571.1 |
- |
6, 7 |
711 |
|
AK315160.1 |
- |
6, 7 |
931 |
|
BC012559.1 |
- |
6, 7 |
948 |
|
BX538002.1 |
- |
5, 6 |
724 |
|
View Full Product Details