Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018948.3 | NP_061821.1 | 4 | 1459 | |
XM_005263477.2 | XP_005263534.1 | 4 | 1473 | ||
XM_006710697.2 | XP_006710760.1 | 4 | 2643 | ||
XM_011541596.2 | XP_011539898.1 | 3 | 1380 | ||
GenBank mRNA | AJ276373.1 | - | 3 | 1210 | |
AK027830.1 | - | 1 | 554 | ||
AK096149.1 | - | 4 | 1434 | ||
AK315718.1 | - | 4 | 1414 | ||
AL137274.1 | - | 1 | 308 | ||
BC025337.1 | - | 4 | 1422 | 4810371 |