Gene Transcripts
- Gene Symbol
- MIER2
- Entrez Gene ID
- 54531
- Gene Name
- MIER family member 2
- Gene Aliases
- KIAA1193, Mi-er2
- Chromosome Location
- Chr. 19: 305573 - 344793 on Build GRCh38
- UniGene ID
- Hs.101891
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017550.1 |
NP_060020.1
|
4, 5 |
387 |
|
XM_005259582.4 |
XP_005259639.1
|
4, 5 |
971 |
|
XM_006722769.3 |
XP_006722832.1
|
4, 5 |
975 |
|
XM_011528072.2 |
XP_011526374.1
|
4, 5 |
973 |
|
XM_011528073.1 |
XP_011526375.1
|
4, 5 |
389 |
|
XM_011528074.2 |
XP_011526376.1
|
4, 5 |
977 |
|
XM_011528075.2 |
XP_011526377.1
|
4, 5 |
1494 |
|
XM_011528077.2 |
XP_011526379.1
|
5, 6 |
3335 |
|
XM_011528081.2 |
XP_011526383.1
|
2, 3 |
231 |
|
XM_011528082.2 |
XP_011526384.1
|
2, 3 |
162 |
|
GenBank mRNA |
AB033019.1 |
- |
|
375 |
|
BC028203.1 |
- |
4, 5 |
387 |
5201526 |
View Full Product Details