Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127458.1 | NP_001120930.1 | 4 | 452 | |
NM_001323561.1 | NP_001310490.1 | 4 | 460 | ||
NM_001323562.1 | NP_001310491.1 | 4 | 467 | ||
NM_001323563.1 | NP_001310492.1 | 4 | 835 | ||
NM_001323564.1 | NP_001310493.1 | 4 | 461 | ||
NM_019095.5 | NP_061968.1 | 4 | 773 | ||
XM_005260738.4 | XP_005260795.1 | 4 | 437 | ||
XM_011529263.1 | XP_011527565.1 | 5 | 791 | ||
GenBank mRNA | AF241784.1 | - | 4 | 692 | |
AL832419.1 | - | 4 | 520 | ||
BM563835.1 | - | 4 | 455 | ||
BP368984.1 | - | 4 | 452 | ||
BU852955.1 | - | 5 | 480 | ||
DQ386730.1 | - | 4 | 599 |