Gene Transcripts

Gene Symbol
MARCH5
Entrez Gene ID
54708
Gene Name
membrane associated ring-CH-type finger 5
Gene Aliases
MARCH-V, MITOL, RNF153
Chromosome Location
Chr. 10: 92291163 - 92353964 on Build GRCh38
UniGene ID
Hs.573490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017824.4 NP_060294.1 5 1029
XM_011539886.2 XP_011538188.1 6 1177
GenBank mRNA AB191202.1 - 5 785
AK000452.1 - 5 1031
AK294799.1 - 5 1103
AK309175.1 - 4 898
BC015480.1 - 5 989 3905766

View Full Product Details