Gene Transcripts

Gene Symbol
RNF111
Entrez Gene ID
54778
Gene Name
ring finger protein 111
Gene Aliases
ARK
Chromosome Location
Chr. 15: 58987588 - 59097419 on Build GRCh38
UniGene ID
Hs.404423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270528.1 NP_001257457.1 2 1004
NM_001270529.1 NP_001257458.1 2 1004
NM_001270530.1 NP_001257459.1 2 861
NM_017610.7 NP_060080.6 2 1004
XM_005254479.3 XP_005254536.1 2 1044
XM_006720575.3 XP_006720638.1 3 1215
XM_006720579.3 XP_006720642.1 2 861
XM_011521694.2 XP_011519996.2 2 997
XM_011521696.2 XP_011519998.2 2 691
XM_011521697.2 XP_011519999.2 2 691
XM_011521699.2 XP_011520001.2 2 686
XM_017022336.1 XP_016877825.1 3 1134
XM_017022337.1 XP_016877826.1 2 1044
XM_017022338.1 XP_016877827.1 2 1082
XM_017022339.1 XP_016877828.1 2 1044
XM_017022340.1 XP_016877829.1 2 1044
XM_017022341.1 XP_016877830.1 2 861
XM_017022342.1 XP_016877831.1 2 858
XM_017022343.1 XP_016877832.1 2 861
GenBank mRNA AK131304.1 - 2 862
BC060862.1 - 2 1007 30336202
BX538130.1 - 2 1005
BX647259.1 - 2 718

View Full Product Details