Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000735.3 | NP_000726.1 | 3 | 328 | |
NM_001252383.1 | NP_001239312.1 | 4 | 421 | ||
GenBank mRNA | AK311972.1 | - | 3 | 285 | |
BC010957.1 | - | 3 | 291 | 4043527 | |
BC020782.1 | - | 3 | 287 | 4732686 | |
BC055080.1 | - | 3 | 289 | 4044086 | |
BG571584.1 | - | 4 | 378 | ||
BG619154.1 | - | 3 | 329 | ||
CR457427.1 | - | 2 | 186 | ||
FJ624078.1 | - | 2 | 114 | ||
FJ624079.1 | - | 2 | 186 | ||
V00518.1 | - | 3 | 236 |