Gene Transcripts

Gene Symbol
ZCCHC10
Entrez Gene ID
54819
Gene Name
zinc finger CCHC-type containing 10
Gene Aliases
Chromosome Location
Chr. 5: 132996985 - 133026621 on Build GRCh38
UniGene ID
Hs.29700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300816.2 NP_001287745.1 3 198
NM_001300817.2 NP_001287746.1 3 198
NM_001300818.2 NP_001287747.1 3 198
NM_001300819.2 NP_001287748.1 2 132
NM_001300822.2 NP_001287751.1 3 198
NM_001308127.1 NP_001295056.1 3 228
NM_001308130.1 NP_001295059.1 2 132
NM_017665.3 NP_060135.1 2 132
NR_125337.2 2 136
NR_131766.1 3 253
NR_131767.1 2 136
XM_011543502.1 XP_011541804.1 3 227
XM_011543503.2 XP_011541805.1 3 245
GenBank mRNA AF086446.1 - 1 14
AK000101.1 - 2 76
AK122970.1 - 3 143
AK300543.1 - 2 100
BC005211.1 - 2 89 3961163
BC022443.1 - 3 139 4285562
BF983215.1 - 2 66
BG699335.1 - 3 178
BI826979.1 - 2 132
BM470321.1 - 2 83
BU165290.1 - 3 129
CB112418.1 - 3 139
DA466245.1 - 3 194
DA528172.1 - 2 76
DA905071.1 - 3 173

View Full Product Details