Gene Transcripts

Gene Symbol
RASIP1
Entrez Gene ID
54922
Gene Name
Ras interacting protein 1
Gene Aliases
RAIN
Chromosome Location
Chr. 19: 48720585 - 48740833 on Build GRCh38
UniGene ID
Hs.233955
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017805.2 NP_060275.2 10 2606
XM_011527053.2 XP_011525355.2 10 2726
XM_011527054.2 XP_011525356.2 10 2726
XM_011527055.2 XP_011525357.2 7 2432
XM_017026914.1 XP_016882403.1 10 2726
XM_017026915.1 XP_016882404.1 7 2432
GenBank mRNA AK000408.1 - 6 721
AK123179.1 - 2 903
AY378097.1 - 10 2471
BC021860.1 - 6 736
BC028614.1 - 10 2478 4815880
BX389914.2 - 2 105

View Full Product Details