Gene Transcripts
- Gene Symbol
- C20orf27
- Entrez Gene ID
- 54976
- Gene Name
- chromosome 20 open reading frame 27
- Gene Aliases
- Chromosome Location
- Chr. 20: 3753499 - 3768388 on Build GRCh38
- UniGene ID
- Hs.274422
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039140.2 |
NP_001034229.1
|
3, 4 |
458 |
|
NM_001258429.1 |
NP_001245358.1
|
3, 4 |
383 |
|
NM_001258430.1 |
NP_001245359.1
|
3, 4 |
837 |
|
NR_047675.1 |
|
3, 4 |
527 |
|
XM_011529266.2 |
XP_011527568.1
|
3, 4 |
438 |
|
GenBank mRNA |
AK000557.1 |
- |
3, 4 |
322 |
|
AK024220.1 |
- |
3, 4 |
322 |
|
AK097645.1 |
- |
3, 4 |
965 |
|
AK124341.1 |
- |
3, 4 |
388 |
|
AK125202.1 |
- |
1, 2 |
1291 |
|
AK225109.1 |
- |
3, 4 |
322 |
|
AK290955.1 |
- |
3, 4 |
527 |
|
AK308520.1 |
- |
3, 4 |
322 |
|
BC012196.1 |
- |
3, 4 |
309 |
2822218 |
BC024036.1 |
- |
3, 4 |
320 |
3510659 |
BF978883.1 |
- |
3, 4 |
384 |
|
BG474804.1 |
- |
3, 4 |
377 |
|
View Full Product Details