Gene Transcripts
- Gene Symbol
- BANK1
- Entrez Gene ID
- 55024
- Gene Name
- B-cell scaffold protein with ankyrin repeats 1
- Gene Aliases
- BANK
- Chromosome Location
- Chr. 4: 101790482 - 102074812 on Build GRCh38
- UniGene ID
- Hs.480400
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001083907.2 |
NP_001077376.2
|
7, 8 |
1245 |
|
NM_001127507.2 |
NP_001120979.2
|
6, 7 |
1077 |
|
NM_017935.4 |
NP_060405.4
|
7, 8 |
1476 |
|
XM_017008337.1 |
XP_016863826.1
|
7, 8 |
1223 |
|
GenBank mRNA |
AB063170.1 |
- |
6, 7 |
1112 |
|
AK000713.1 |
- |
7, 8 |
1192 |
|
AK091523.1 |
- |
11, 12 |
1577 |
|
AK292133.1 |
- |
7, 8 |
1320 |
|
AL834291.1 |
- |
7, 8 |
1295 |
|
BC032241.1 |
- |
7, 8 |
1344 |
5214147 |
EU051376.1 |
- |
6, 7 |
815 |
|
View Full Product Details