Gene Transcripts

Gene Symbol
MSTO1
Entrez Gene ID
55154
Gene Name
misato 1, mitochondrial distribution and morphology regulator
Gene Aliases
LST005, MST
Chromosome Location
Chr. 1: 155610170 - 155614967 on Build GRCh38
UniGene ID
Hs.656547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256532.1 NP_001243461.1 6 528
NM_001256533.1 NP_001243462.1 6 528
NM_018116.3 NP_060586.2 6 528
NR_046292.1 6 650
NR_046293.1 6 592
NR_046295.1 5 452
XM_006711421.1 XP_006711484.1 5 487
XM_006711423.2 XP_006711486.1 6 536
XM_011509695.1 XP_011507997.1 6 486
XM_011509696.1 XP_011507998.1 6 486
XM_011509697.1 XP_011507999.1 6 486
XM_011509698.1 XP_011508000.1 6 486
XM_011509699.1 XP_011508001.1 6 486
XM_011509700.1 XP_011508002.1 6 486
XM_011509701.1 XP_011508003.1 6 486
XM_011509702.1 XP_011508004.1 6 486
XM_011509703.1 XP_011508005.1 5 1128
XM_011509704.1 XP_011508006.1 6 1051
XM_011509705.1 XP_011508007.1 5 1065
XM_011509706.1 XP_011508008.1 6 460
XM_011509707.1 XP_011508009.1 6 993
XM_011509708.1 XP_011508010.1 6 929
XM_011509709.2 XP_011508011.1 6 486
XM_017001603.1 XP_016857092.1 6 524
XM_017001604.1 XP_016857093.1 6 462
XM_017001605.1 XP_016857094.1 6 993
XM_017001606.1 XP_016857095.1 6 929
XM_017001607.1 XP_016857096.1 6 1067
XM_017001608.1 XP_016857097.1 5 514
XM_017001609.1 XP_016857098.1 6 536
XM_017001610.1 XP_016857099.1 6 462
XM_017001611.1 XP_016857100.1 6 993
XM_017001612.1 XP_016857101.1 5 429
XM_017001613.1 XP_016857102.1 5 365
XM_017001614.1 XP_016857103.1 6 536
XM_017001615.1 XP_016857104.1 6 536
XM_017001616.1 XP_016857105.1 2 205
GenBank mRNA AF111708.1 - 3 155
AF272833.1 - 6 406
AK001366.1 - 6 457
AK092950.1 - 5 352
AK222863.1 - 6 454
AK290610.1 - 6 503
AK297140.1 - 6 579
AK309573.1 - 6 513
AY334564.1 - 5 571
BC002535.2 - 6 473 3140498
BC070067.1 - 6 544
BX537684.1 - 5 404
CR749791.1 - 6 454
CR936872.1 - 3 1341

Gene Symbol
MSTO2P
Entrez Gene ID
100129405
Gene Name
misato family member 2, pseudogene
Gene Aliases
MSTO2
Chromosome Location
Chr. 1: 155745768 - 155750688 on Build GRCh38
UniGene ID
Hs.656361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024117.2 6 574
GenBank mRNA AK293928.1 - 6 510
AK295146.1 - 6 574
BC099909.1 - 6 524

View Full Product Details