Gene Transcripts

Gene Symbol
MSL2
Entrez Gene ID
55167
Gene Name
male-specific lethal 2 homolog (Drosophila)
Gene Aliases
MSL-2, MSL2L1, RNF184
Chromosome Location
Chr. 3: 136148918 - 136197300 on Build GRCh38
UniGene ID
Hs.18631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145417.1 NP_001138889.1 2 907
NM_018133.3 NP_060603.2 2 1708
XM_005247571.3 XP_005247628.1 2 1442
XM_006713684.3 XP_006713747.1 2 925
XM_011512949.2 XP_011511251.1 2 1012
GenBank mRNA AB046805.1 - 2 1056
AK091956.1 - 2 1708
AK292557.1 - 2 1203
AK302491.1 - 2 874
AL834289.1 - 2 1026
BC032719.1 - 1 497
BC093764.1 - 2 1008 7939609
BC093790.1 - 2 1007 7939635

View Full Product Details