Gene Transcripts

Gene Symbol
SBNO1
Entrez Gene ID
55206
Gene Name
strawberry notch homolog 1
Gene Aliases
MOP3, Sno
Chromosome Location
Chr. 12: 123289109 - 123365204 on Build GRCh38
UniGene ID
Hs.7012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167856.1 NP_001161328.1 20, 21 2942
NM_018183.3 NP_060653.3 20, 21 2939
XM_005253572.3 XP_005253629.1 21, 22 3080
XM_005253573.4 XP_005253630.1 22, 23 3490
XM_005253575.3 XP_005253632.1 21, 22 3077
XM_005253576.3 XP_005253633.1 22, 23 3121
XM_006719473.3 XP_006719536.1 23, 24 3246
XM_006719474.3 XP_006719537.1 22, 23 3199
XM_011538533.2 XP_011536835.1 23, 24 3240
XM_011538534.2 XP_011536836.1 20, 21 2978
XM_011538535.2 XP_011536837.1 20, 21 2980
XM_017019556.1 XP_016875045.1 21, 22 3064
XM_017019557.1 XP_016875046.1 21, 22 3071
XM_017019558.1 XP_016875047.1 22, 23 3134
XM_017019559.1 XP_016875048.1 21, 22 3201
GenBank mRNA AB014772.1 - 20, 21 2939
AK074256.1 - 4, 5 509
AK096864.1 - 3, 4 312
BC133704.1 - 20, 21 2942 40124902

View Full Product Details