Gene Transcripts

Gene Symbol
UBA6
Entrez Gene ID
55236
Gene Name
ubiquitin like modifier activating enzyme 6
Gene Aliases
E1-L2, MOP-4, UBE1L2
Chromosome Location
Chr. 4: 67615761 - 67701171 on Build GRCh38
UniGene ID
Hs.212774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018227.5 NP_060697.4 10 917
XM_017008359.1 XP_016863848.1 10 894
GenBank mRNA AB014773.1 - 10 865
AK094164.1 - 2 123
AK094969.1 - 10 894
AK314371.1 - 10 894
AL832015.2 - 10 886
AL832458.2 - 10 886
BC031637.1 - 10 899
BC126484.1 - 10 912 8992200
BC126486.1 - 10 912 8992202
EF623993.1 - 10 865

View Full Product Details