Gene Transcripts

Gene Symbol
UFSP2
Entrez Gene ID
55325
Gene Name
UFM1 specific peptidase 2
Gene Aliases
BHD, C4orf20
Chromosome Location
Chr. 4: 185399540 - 185425985 on Build GRCh38
UniGene ID
Hs.713548
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018359.3 NP_060829.2 5 548
NR_028085.1 5 523
GenBank mRNA AK002062.1 - 5 477
AK091635.1 - 5 516
BC010493.1 - 4 444 3048997
BP264462.1 - 5 544
CR457278.1 - 4 431
DA537391.1 - 5 542

View Full Product Details