Gene Transcripts

Gene Symbol
NCBP3
Entrez Gene ID
55421
Gene Name
nuclear cap binding subunit 3
Gene Aliases
C17orf85, ELG, HSA277841
Chromosome Location
Chr. 17: 3806748 - 3846251 on Build GRCh38
UniGene ID
Hs.120963
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114118.2 NP_001107590.1 13 1821
XM_017024826.1 XP_016880315.1 12 1708
XM_017024829.1 XP_016880318.1 8 1113
GenBank mRNA AJ277841.1 - 8 1118
AK125048.1 - 10 2510
AK223470.1 - 6 997
BC010707.1 - 6 936

View Full Product Details