Gene Transcripts

Gene Symbol
PNRC2
Entrez Gene ID
55629
Gene Name
proline rich nuclear receptor coactivator 2
Gene Aliases
Chromosome Location
Chr. 1: 23959164 - 23963462 on Build GRCh38
UniGene ID
Hs.512636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017761.3 NP_060231.1 3 759
XM_017001691.1 XP_016857180.1 3 664
GenBank mRNA AF151042.1 - 3 743
AF374386.1 - 3 677
AK000319.1 - 2 480
AK024774.1 - 1 179
AK300522.1 - 3 500
AK311305.1 - 4 544
BC001959.1 - 3 730 3536977
BC078177.1 - 3 711 6526331
BC085018.1 - 3 729 6650445

View Full Product Details